Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRC1 Monoclonal Antibody (1135)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA123291
Description
Recommended positive controls: mouse brain.
Clone 1135 recognizes SRC-1, steroid-receptor coactivator-1. SRC-1 is a member of a gene family that functions in the full transcriptional activation of the steroid hormone receptor superfamily and contains intrinsic histone acetyltransferase activity. SRC-1 has a molecular weight of 160 kDa and has a basic helix-loop-helix (bHLH)-PAS domain.
Specifications
| SRC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P70365, Q15788 | |
| NCOA1 | |
| Amino acids 477-947 of human SRC-1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG1 |
| ChIP Assay, Immunoprecipitation, Western Blot | |
| 1135 | |
| Unconjugated | |
| NCOA1 | |
| ASV; bHLHe42; bHLHe74; class E basic helix-loop-helix protein 74; c-SRC; F-SRC-1; Hin-2 protein; KAT13A; mNRC-1; NCOA1; NCoA-1; Nuclear receptor coactivator 1; nuclear receptor coactivator protein 1; p60-Src; PAX3/NCOA1 fusion protein; Protein Hin-2; Renal carcinoma antigen NY-REN-52; RIP160; Src1; SRC-1; SRC-1 (-Q); SRC-a/NCoA-1; Steroid receptor coactivator 1; steroid receptor coactivator-1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 17977, 313929, 8648 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction