Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRC1 Rabbit anti-Human, Clone: 3S4C6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | SRC1 |
---|---|
Clone | 3S4C6 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
SRC1 Monoclonal antibody specifically detects SRC1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
SRC1 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Chromatin Research, Transcription Factors and Regulators | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
8648 | |
IgG | |
Affinity purified |
3S4C6 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
bHLHe42, BHLHE74, Class E basic helix-loop-helix protein 74, EC 2.3.1.48, Hin-2 protein, KAT13A, MGC129719, NCoA-1, nuclear receptor coactivator 1, PAX3/NCOA1 fusion protein, Protein Hin-2, Renal carcinoma antigen NY-REN-52, RIP160bHLHe74F-SRC-1, SRC-1, SRC1MGC129720, Steroid receptor coactivator 1, steroid receptor coactivator-1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 400-750 of human SRC1 (Q15788). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title