Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    SRC1 Rabbit anti-Human, Clone: JB66-31, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275672
Description
SRC1 Monoclonal specifically detects SRC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SRC1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:100 | |
| bHLHe42, BHLHE74, Class E basic helix-loop-helix protein 74, EC 2.3.1.48, Hin-2 protein, KAT13A, MGC129719, NCoA-1, nuclear receptor coactivator 1, PAX3/NCOA1 fusion protein, Protein Hin-2, Renal carcinoma antigen NY-REN-52, RIP160bHLHe74F-SRC-1, SRC-1, SRC1MGC129720, Steroid receptor coactivator 1, steroid receptor coactivator-1 | |
| Rabbit | |
| 157 kDa | |
| 100 μL | |
| Primary | |
| Human | |
| Purified | 
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| JB66-31 | |
| Unconjugated | |
| TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
| NCOA1 | |
| Recombinant protein within Human SRC1 aa 380-770/ 1,441. (SwissProt: Q15788 Human) | |
| Protein A purified | |
| Cancer, Chromatin Research, Transcription Factors and Regulators | |
| 8648.0 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            -Immunohistochemistry-Paraffin-NBP2-75672-img0003.jpg-250.jpg) 
                            -Immunohistochemistry-Paraffin-NBP2-75672-img0002.jpg-250.jpg) 
                            -Western Blot-NBP2-75672-img0004.jpg-250.jpg) 
                            -Immunohistochemistry-Paraffin-NBP2-75672-img0001.jpg-250.jpg)