Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRC1 Recombinant Rabbit Monoclonal Antibody (3S4C6)
GREENER_CHOICE

Catalog No. PIMA542905
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542905 Supplier Invitrogen™ Supplier No. MA542905
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HeLa, 293T, SGC-7901. Immunogen sequence: VARSSTLPPS NSNMVSTRIN RQQSSDLHSS SHSNSSNSQG SFGCSPGSQI VANVALNQGQ ASSQSSNPSL NLNNSPMEGT GISLAQFMSP RRQVTSGLAT RPRMPNNSFP PNISTLSSPV GMTSSACNNN NRSYSNIPVT SLQGMNEGPN NSVGFSASSP VLRQMSSQNS PSRLNIQPAK AESKDNKEIA SILNEMIQSD NSSSDGKPLD SGLLHNNDRL SDGDSKYSQT SHKLVQLLTT TAEQQLRHAD IDTSCKDVLS CTGTSNSASA NSSGGSCPSS HSSLTERHKI LHRLLQEGSP SDITTLSVEP DKKDSASTSV SVTGQVQGNS SIKLELDASK KKESKDHQLL R.

Clone 1135 recognizes SRC-1, steroid-receptor coactivator-1. SRC-1 is a member of a gene family that functions in the full transcriptional activation of the steroid hormone receptor superfamily and contains intrinsic histone acetyltransferase activity. SRC-1 has a molecular weight of 160 kDa and has a basic helix-loop-helix (bHLH)-PAS domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRC1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 3S4C6
Concentration 1.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NCOA1
Gene Accession No. Q15788
Gene Alias ASV; bHLHe42; bHLHe74; class E basic helix-loop-helix protein 74; c-SRC; F-SRC-1; Hin-2 protein; KAT13A; mNRC-1; NCOA1; NCoA-1; Nuclear receptor coactivator 1; nuclear receptor coactivator protein 1; p60-Src; PAX3/NCOA1 fusion protein; Protein Hin-2; Renal carcinoma antigen NY-REN-52; RIP160; Src1; SRC-1; SRC-1 (-Q); SRC-a/NCoA-1; Steroid receptor coactivator 1; steroid receptor coactivator-1
Gene Symbols NCOA1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 400-750 of human KAT13A / SRC1 (Q15788).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8648
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.