Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120079 Supplier Invitrogen™ Supplier No. PA5120079
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, BT-474. The target is usually found in the following locations: Nucleus. Immunogen sequence: SSLASPDLKM GNLQNSPVNM NPPPLSKMGS LDSKDCFGLY GEPSEGTTGQ AESSCHPGEQ KETNDPNLPP AVSSERADGQ SRLHDSKGQT KLLQLLTTKS DQMEPSPLAS SLSDTNKDST GSLPGSGSTH GTSLKEKHKI LHRLLQDSSS PVDLAKLTAE ATGKDLSQES SSTAPGSEVT IKQEPVSPKK KENALLRYLL DKDDTKDIGL PEITPKLERL DSKTDPASNT KLIAMKTEKE EMSFEPGDQP GSELDNLEEI LDDLQNSQLP Q.

SRC2 (steroid receptor coactivator 2), also known as nuclear receptor coactivator2, NCOA2 or TIF2, aids in the function of nuclear hormone receptors. Nuclear hormone receptors are conditional transcription factors that play important roles in various aspects of cell growth, development, and homeostasis by controlling expression of specific genes. Members of the nuclear hormone receptor superfamily, which includes the 5 steroid receptors and class II nuclear receptors, are structurally characterized by 3 distinct domains: an N-terminal transcriptional activation domain, a central DNA-binding domain, and a C-terminal hormone-binding domain. Before the binding of hormone, steroid receptors, which are sometimes called class I of the nuclear hormone receptor family, remain inactive in a complex with heat-shock protein-90 and other stress family proteins. Binding of hormone induces critical conformational changes in steroid receptors that cause them to dissociate from the inhibitory complex, bind as homodimers to specific DNA enhancer elements associated with target genes, and modulate that gene's transcription. After binding to enhancer elements, transcription factors require transcriptional coactivator proteins to mediate their stimulation of transcription initiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRC2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NCOA2
Gene Accession No. Q15596, Q9WUI9
Gene Alias bHLHe75; c-fgr; Class E basic helix-loop-helix protein 75; c-src2; D1Ertd433e; FLJ43153; Glucocorticoid receptor-interacting protein 1; glucocorticoid receptor-interacting protein-1; Grip1; GRIP-1; hTIF2; I79_019839; KAT13C; MGC75096; NCOA2; NCoA-2; Nuclear receptor coactivator 2; p160 steroid receptor coactivator 2; p55c-fgr; p58c-fgr; SRC2; SRC-2; Steroid receptor coactivator 2; TIF 2; Tif2; transcriptional intermediary factor 2
Gene Symbols NCOA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 550-820 of human NCOA2 (NP_006531.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10499, 83724
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.