Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRCAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP184063
Description
SRCAP Polyclonal specifically detects SRCAP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SRCAP | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
domino homolog 1, Domino homolog 2, DOMO1, EAF1, EC 3.6.1, EC 3.6.4.-, FLJ44499, KIAA0309helicase SRCAP, Snf2-related CBP activator, Snf2-related CREBBP activator protein, Swi2/Snf2-related ATPase homolog, domino homolog 1, SWR1Snf2-related CBP activator protein | |
Rabbit | |
Affinity Purified | |
RUO | |
10847 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
SRCAP | |
This antibody was developed against Recombinant Protein corresponding to amino acids:AQALASPESLELASVASSETSSLSLVPPKDLLPVAVEILPVSEKNLSLTPSAPSLTLEAGSIPNGQEQEAPDSAEGTTLTVLPEGEELPLCVSESNGLELPPSAASDEPLQEPLEADRTSEELTEAKTPTSS | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction