Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRCAP Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537921
Description
This antibody is predicted to react with mouse, rat, dog and pig based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

Specifications
SRCAP | |
Polyclonal | |
Unconjugated | |
SRCAP | |
B930091H02Rik; D030022P06Rik; domino homolog 2; DOMO1; EAF1; F630004O05Rik; FLHS; FLJ44499; helicase SRCAP; KIAA0309; RGD1565642; Snf2-related CBP activator; Snf2-related CBP activator protein; Snf2-related CREBBP activator protein; SRCAP; Swi2/Snf2-related ATPase homolog, domino homolog 1; SWR1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
10847 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q6ZRS2 | |
SRCAP | |
Peptide with sequence C-QEGNDAEAQRREIE, from the internal region near N-terminus of SRCAP (aa309-322). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction