Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRCRB4D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116269
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116269 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116269 Supplier Invitrogen™ Supplier No. PA5116269
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRCRB4D Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SRCRB4D
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SSC4D
Gene Accession No. Q8WTU2
Gene Alias Four scavenger receptor cysteine-rich domains-containing protein; S4D-SRCRB; S4-SRCRB; scavenger receptor cysteine rich domain containing, group B (4 domains); scavenger receptor cysteine rich family member with 4 domains; scavenger receptor cysteine rich family, 4 domains; scavenger receptor cysteine-rich domain-containing group B protein; scavenger receptor cysteine-rich protein SRCRB-S4D; SRCRB4D; SRCRB-S4D; SSC4D
Gene Symbols SSC4D
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 165-191 amino acids from the Central region of human SRCRB4D.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136853
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.