Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRCRB4D Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118284
This item is not returnable.
View return policy
Description
Specificity: Human SRCRB4D.
Specifications
| SRCRB4D | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q8WTU2 | |
| SSC4D | |
| E. coli-derived Human SSC4D fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| SSC4D | |
| Four scavenger receptor cysteine-rich domains-containing protein; S4D-SRCRB; S4-SRCRB; scavenger receptor cysteine rich domain containing, group B (4 domains); scavenger receptor cysteine rich family member with 4 domains; scavenger receptor cysteine rich family, 4 domains; scavenger receptor cysteine-rich domain-containing group B protein; scavenger receptor cysteine-rich protein SRCRB-S4D; SRCRB4D; SRCRB-S4D; SSC4D | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 136853 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction