Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRcyp Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501561710
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-1710 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-1710 Supplier Bethyl Laboratories, Inc Supplier No. A302075AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

PPIG belongs to a highly conserved class of cyclophilins that function as peptidyl-prolyl-isomerases (PPIases) to catalyze the conversion of cis-proline to trans-proline in a polypeptide chain. PPIG contains an amino-terminal cyclophilin domain followed by Nopp140 repeats that are involved in its function as a nuclear chaperone. The carboxy-terminal of PPIG contains a SR (arginine-serine dipeptide repeat) domain that is involved in pre-mRNA splicing and processing. PPIG interacts with the carboxy-terminal domain of RNA polymerase II as well as several other SR family splicing factors. These interactions lead to changes in localization and conformation and suggest a regulatory role in transcription and pre-mRNA splicing in the elongating RNA polymerase complex. PPIG is found in the nuclear matrix and nuclear speckles and is involved in the regulation of gene expression. PPIG shows a predomitly diffuse cytoplasmic distribution at the onset of mitosis, and in late telophase the isomerase is recruited to the newly formed nuclei.

Specifications

Antigen SRcyp
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PPIG
Gene Accession No. Q13427
Gene Alias CARS-cyclophilin; CASP10; Clk-associating RS-cyclophilin; OTTHUMP00000204761; OTTHUMP00000204762; PPIase G; SR-cyclophilin; SR-cyp; SR-related CTD-associated factor 10; cyclophilin G; peptidyl-prolyl cis-trans isomerase G; peptidyl-prolyl isomerase G (cyclophilin G); rotamase G
Gene Symbols PPIG
Host Species Rabbit
Immunogen Between 375 and 325
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9360
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.