Learn More
SRcyp Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302076A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
PPIG belongs to a highly conserved class of cyclophilins that function as peptidyl-prolyl-isomerases (PPIases) to catalyze the conversion of cis-proline to trans-proline in a polypeptide chain. PPIG contains an amino-terminal cyclophilin domain followed by Nopp140 repeats that are involved in its function as a nuclear chaperone. The carboxy-terminal of PPIG contains a SR (arginine-serine dipeptide repeat) domain that is involved in pre-mRNA splicing and processing. PPIG interacts with the carboxy-terminal domain of RNA polymerase II as well as several other SR family splicing factors. These interactions lead to changes in localization and conformation and suggest a regulatory role in transcription and pre-mRNA splicing in the elongating RNA polymerase complex. PPIG is found in the nuclear matrix and nuclear speckles and is involved in the regulation of gene expression. PPIG shows a predomitly diffuse cytoplasmic distribution at the onset of mitosis, and in late telophase the isomerase is recruited to the newly formed nuclei.Specifications
SRcyp | |
Polyclonal | |
Unconjugated | |
PPIG | |
CARS-cyclophilin; CASP10; Clk-associating RS-cyclophilin; OTTHUMP00000204761; OTTHUMP00000204762; PPIase G; SR-cyclophilin; SR-cyp; SR-related CTD-associated factor 10; cyclophilin G; peptidyl-prolyl cis-trans isomerase G; peptidyl-prolyl isomerase G (cyclophilin G); rotamase G | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9360 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q13427 | |
PPIG | |
Between 450 and 500 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.