Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRD5A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519091
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519091 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519091 Supplier Invitrogen™ Supplier No. PA519091
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

This gene encodes a microsomal protein expressed at high levels in androgen-sensitive tissues such as the prostate. The encoded protein is active at acidic pH and is sensitive to the 4-azasteroid inhibitor finasteride. Deficiencies in this gene can result in male pseudohermaphroditism, specifically pseudovaginal perineoscrotal hypospadias (PPSH).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRD5A2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SRD5A2
Gene Accession No. P31213
Gene Alias 3-oxo-5-alpha-steroid 4-dehydrogenase 2; 5 alpha-SR2; 5ART2; MGC138457; S5AR 2; SR type 2; SRD5A2; ST5AR2; steroid 5 alpha reductase type2; steroid 5 alpha-reductase 2; steroid 5-alpha-reductase 2; steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2); type 2; 4-ene-3-oxosteroid 5-alpha oxidoreductase; type II 5-alpha reductase
Gene Symbols SRD5A2
Host Species Goat
Immunogen Synthetic peptide sequence (KMFEDYPKSRK) corresponding to the C-terminus amino acids of SRD5A2 (aa 237-248).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6716
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.