Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRD5A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525465
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525465 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525465 Supplier Invitrogen™ Supplier No. PA525465
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRD5A2 Polyclonal Antibody for Western Blot

This gene encodes a microsomal protein expressed at high levels in androgen-sensitive tissues such as the prostate. The encoded protein is active at acidic pH and is sensitive to the 4-azasteroid inhibitor finasteride. Deficiencies in this gene can result in male pseudohermaphroditism, specifically pseudovaginal perineoscrotal hypospadias (PPSH).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRD5A2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SRD5A2
Gene Accession No. P31213, Q99N99
Gene Alias 3-oxo-5-alpha-steroid 4-dehydrogenase 2; 5 alpha-SR2; 5ART2; MGC138457; S5AR 2; SR type 2; SRD5A2; ST5AR2; steroid 5 alpha reductase type2; steroid 5 alpha-reductase 2; steroid 5-alpha-reductase 2; steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2); type 2; 4-ene-3-oxosteroid 5-alpha oxidoreductase; type II 5-alpha reductase
Gene Symbols SRD5A2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 153-182 amino acids from the central region of human SRD5A2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6716, 94224
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.