Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRD5A2 Rabbit anti-Human, Mouse, Rat, Clone: 8A4H5, Novus Biologicals™
SDP

Catalog No. p-7247397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166896 20 μg
NB166895 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166896 Supplier Novus Biologicals Supplier No. NBP31590020UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SRD5A2 Monoclonal antibody specifically detects SRD5A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen SRD5A2
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 8A4H5
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 3-oxo-5-alpha-steroid 4-dehydrogenase 2,5 alpha-SR2, EC 1.3.99.5, MGC138457, S5AR 2, SR type 2, Steroid 5-alpha-reductase 2,3-oxo-5 alpha-steroid 4-dehydrogenase 2, steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta4-dehydrogenase alpha 2), Type II 5-alpha reductase
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SRD5A2 (P31213). MQVQCQQSPVLAGSATLVALGALALYVAKPSGYGKHTESLKPAATRLPARAAWFLQELPSFAVPAGILARQPLSLFGPPGTVLLGLFCVHYFHRTFVYSL
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6716
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.