Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRD5A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555480
Description
Immunogen sequence: KCHVILGNLR KNKAGVVIHC NHRIPFGDWF EYVSSPNYL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
SRD5A3 belongs to the steroid 5-alpha reductase family and converts testosterone (T) into 5-alpha-dihydrotestosterone (DHT).
Specifications
SRD5A3 | |
Polyclonal | |
Unconjugated | |
SRD5A3 | |
3-oxo-5-alpha-steroid 4-dehydrogenase 3; CDG1P; CDG1Q; KRIZI; polyprenol reductase; probable polyprenol reductase; S5AR 3; SR type 3; SRD5A2L; SRD5A2L1; SRD5A3; steroid 5 alpha-reductase 3; Steroid 5-alpha-reductase 2-like; Steroid 5-alpha-reductase 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79644 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H8P0 | |
SRD5A3 | |
Recombinant protein corresponding to Human SRD5A3. Recombinant protein control fragment (Product #RP-91266). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction