Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SREB3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NLS516
Description
SREB3 Polyclonal specifically detects SREB3 in Human, Bat, Canine, Monkey, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SREB3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.1% Sodium Azide | |
GPR173 | |
Synthetic 19 amino acid peptide from N-terminal extracellular domain of human SREB3. | |
0.05 mL | |
GPCR | |
54328 | |
Human, Bat, Canine, Monkey, Rabbit | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Immunohistochemistry, Immunohistochemistry-Paraffin 6.4 ug/ml | |
G protein coupled receptor 173, G protein-coupled receptor 173, G-protein coupled receptor 173, probable G-protein coupled receptor 173, SREB3Super conserved receptor expressed in brain 3 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
SREB3 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction