Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SREBP-1/2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS1402R

Description
Sterol regulatory element-binding proteins (SREBPs) are transcription factors that are members of the basic helix-loop-helix leucine zipper family of DNA binding proteins. Three isoforms have been identified in mammalian tissues that vary in structure, regulation, and function. SREBP-1a and SREBP-1c (originally cloned as ADD1) are protein products of alternative promoter usage of the SREBP-1 gene. The third isoform is transcribed from a different gene, SREBP-2. SREBPs are present as 120 kDa inactive precursors in the endoplasmic reticulum (ER) membrane. Upon activation, the SREBP protein is translocated to the Golgi and proteolytic cleavage occurs resulting in a mature transcriptionally active 60-78 kDa fragment. In liver and adipose tissues, SREBPs have a significant influence on lipid and cholesterol accumulation by inducing the transcription of genes involved in these processes. While SREBP-1 is thought to be more important in regulating the expression of genes involved in triglyceride synthesis and accumulation, SREBP-2 has been more closely linked to those involved in cholesterol synthesis and accumulation.Specifications
SREBP-1/2 | |
Polyclonal | |
Unconjugated | |
SREBF1 | |
ADD1; ADD-1; Adipocyte determination- and differentiation-dependent factor 1; BHLHD1; Class D basic helix-loop-helix protein 1; D630008H06; I79_010155; nt-SREBP1; Processed sterol regulatory element-binding protein 1; SREBF1; SREBP1; SREBP-1; SREBP1a; SREBP-1a; SREBP1c; SREBP-1c; sterol regulatory element binding protein 1; sterol regulatory element binding protein-1; sterol regulatory element binding transcription factor 1; sterol regulatory element binding-protein 1; Sterol regulatory element-binding protein 1; Sterol regulatory element-binding transcription factor 1 | |
Rabbit | |
Protein A | |
RUO | |
6720 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P36956 | |
SREBF1 | |
KLH conjugated synthetic peptide derived from human SREBP-1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction