Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP-1 Monoclonal Antibody (IgG-2A4), Active Motif™
SDP

Catalog No. 501992981
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2981 100 μg
50-199-2982 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2981 Supplier Active Motif Supplier No. 39939
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sterol regulatory element-binding proteins (SREBPs) are transcription factors that are members of the basic helix-loop-helix leucine zipper family of DNA binding proteins. Three isoforms have been identified in mammalian tissues that vary in structure, regulation, and function. SREBP-1a and SREBP-1c (originally cloned as ADD1) are protein products of alternative promoter usage of the SREBP-1 gene. The third isoform is transcribed from a different gene, SREBP-2. SREBPs are present as 120 kDa inactive precursors in the endoplasmic reticulum (ER) membrane. Upon activation, the SREBP protein is translocated to the Golgi and proteolytic cleavage occurs resulting in a mature transcriptionally active 60-78 kDa fragment. In liver and adipose tissues, SREBPs have a significant influence on lipid and cholesterol accumulation by inducing the transcription of genes involved in these processes. While SREBP-1 is thought to be more important in regulating the expression of genes involved in triglyceride synthesis and accumulation, SREBP-2 has been more closely linked to those involved in cholesterol synthesis and accumulation.

Specifications

Antigen SREBP-1
Applications ChIP Assay, Gel Shift, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone IgG-2A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene SREBF1
Gene Accession No. P36956
Gene Alias ADD1; ADD-1; Adipocyte determination- and differentiation-dependent factor 1; BHLHD1; Class D basic helix-loop-helix protein 1; D630008H06; I79_010155; nt-SREBP1; Processed sterol regulatory element-binding protein 1; SREBF1; SREBP1; SREBP-1; SREBP1a; SREBP-1a; SREBP1c; SREBP-1c; sterol regulatory element binding protein 1; sterol regulatory element binding protein-1; sterol regulatory element binding transcription factor 1; sterol regulatory element binding-protein 1; Sterol regulatory element-binding protein 1; Sterol regulatory element-binding transcription factor 1
Gene Symbols SREBF1
Host Species Mouse
Immunogen This SREBP-1 antibody was raised against a recombinant protein corresponding to amino acids 301-407 of human SREBP-1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6720
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.