Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP-1, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABS150825UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS150825UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS150825UG Supplier MilliporeSigma Supplier No. ABS150825UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-SREBP-1, Cat. No. ABS1508, is a rabbit polyclonal antibdody that detects Sterol regulatory element-binding protein 1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Sterol regulatory element-binding protein 1 (UniProt: P36956; also known as SREBP-1, Class D basic helix-loop-helix protein 1, bHLHd1, Sterol regulatory element-binding transcription factor 1) is encoded by the SREBF1 (also known as BHLHD1, SREBP1) gene (Gene ID: 6720) in human. SREBP-1 is a multi-pass membrane protein that acts as at transcriptional activator required for lipid homeostasis. Six isoforms of SREBP-1 have been described that are produced by alternative splicing. SREBP-1 is expressed in a wide variety of tissues and is most abundant in liver and adrenal gland. In fetal lung and liver it is expressed in high levels. SREBP-1A predominates in hepatoma cell lines while isoform SREBP-1C is predominantly found in liver, adrenal gland, and ovary. Both isoforms IA and IC is found in kidney, brain, white fat, and muscle. SREBP-1 regulates transcription of the LDL receptor gene as well as the fatty acid and to a lesser degree the cholesterol synthesis pathway. It binds to the sterol regulatory element 1 (SRE-1) (5'-ATCACCCCAC-3') and has dual sequence specificity binding to both an E-box motif (5'-ATCACGTGA-3') and to SRE-1 (5'-ATCACCCCAC-3'). SREBP-1 has two cytoplasmic domains (aa 1-487 and 567-1147; two helical domains (aa 488-508 and 548-568), and a luminal domain (aa 509-547). In the absence of sterols, it moves from the endoplasmic reticulum to the Golgi. At low cholesterol the SCAP/SREBP complex is recruited into COPII vesicles for export from the ER. In the Golgi complex SREBPs are cleaved sequentially by site-1 and site-2 protease. The first cleavage by site-1 protease occurs within the luminal loop, the second cleavage by site-2 protease occurs within the first transmembrane domain and releases the transcription factor from the Golgi membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SREBP-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected SREBP-1 in human adrenal gland and human kidney tissues.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Sterol regulatory element-binding protein 1;Class D basic helix-loop-helix protein 1;bHLHd1;Sterol regulatory element-binding transcription factor 1
Gene Symbols SREBF1;BHLHD1;SREBP1
Host Species Rabbit
Immunogen KLH-conjugated linenar peptide corresponding to 14 amino acids from the N-terminal half of human Sterol regulatory element-binding protein 1. The Immunogen sequence is conserved in SREBP-1A, B, and C and in isoform 4.
Molecular Weight of Antigen ∽120 kDa observed; 121.67 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Sterol regulatory element-binding protein 1 in human.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.