Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SREBP1 Antibody (2A4), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB600582AF750
Description
SREBP1 Monoclonal specifically detects SREBP1 in Human, Mouse, Rat, Canine, Chicken, Hamster, Monkey, Golden Syrian Hamster samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.Specifications
SREBP1 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
SREBF1 | |
6 His-tag fusion protein of human SREBP1 corresponding to amino acids 301-407. [UniProtNo. P36956] | |
Primary | |
Human, Mouse, Rat, Canine, Chicken, Hamster, Monkey, Golden Syrian Hamster | |
IgG1 κ |
Western Blot, Immunohistochemistry, Immunoassay | |
2A4 | |
Western Blot, Immunohistochemistry, Knockout Validated | |
BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1 | |
Mouse | |
0.1 mL | |
6720 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction