Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP1 Antibody (2A4) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$244.50 - $562.50

Specifications

Antigen SREBP1
Clone 2A4
Concentration 1 mg/mL
Dilution Western Blot 1-2μg/mL, Simple Western 1:12.5, Immunohistochemistry-Frozen, Knockout Validated
Applications Western Blot, Immunohistochemistry (Frozen), Immunoassay
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600582SS
SDP
View Documents
Novus Biologicals
NB600582SS
0.025 mL
Each for $244.50
Only null left
Add to Cart
 
NB600582
SDP
View Documents
Novus Biologicals
NB600582
0.1 mL
Each for $562.50
Only null left
Add to Cart
 
Description

Description

SREBP1 Monoclonal specifically detects SREBP1 in Human, Mouse, Rat, Canine, Chicken, Hamster, Monkey, Golden Syrian Hamster samples. It is validated for Western Blot, Simple Western, Immunohistochemistry-Frozen, Knockout Validated.
Specifications

Specifications

SREBP1
1 mg/mL
Western Blot, Immunohistochemistry (Frozen), Immunoassay
Unconjugated
Mouse
Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators
P36956
6720
6 His-tag fusion protein of human SREBP1 corresponding to amino acids 301-407. [UniProt# P36956]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
2A4
Western Blot 1-2μg/mL, Simple Western 1:12.5, Immunohistochemistry-Frozen, Knockout Validated
Monoclonal
Purified
RUO
Human, Mouse, Rat, Canine, Chicken, Hamster, Monkey, Golden Syrian Hamster
BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1
SREBF1
IgG1 κ
Protein G purified
Human, mouse, rat and Golden Syrian Hamster. Canine reactivity reported in scientific literature (PMID: 23720350). Hamster reactivity reported in scientific literature (PMID: 24393244).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.