Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SREBP1 Antibody (SREBP1/1578), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308432FR
Description
SREBP1 Monoclonal antibody specifically detects SREBP1 in Human samples. It is validated for ELISASpecifications
SREBP1 | |
Monoclonal | |
DyLight 680 | |
BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1 | |
Recombinant full-length human SREBP1 protein (Uniprot: P36956) | |
100 μg | |
Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators | |
6720 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA | |
SREBP1/1578 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction