Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SREBP2 Antibody (1D2), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP154446AF350
Description
SREBP2 Monoclonal specifically detects SREBP2 in Human, Chicken samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
SREBP2 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
SREBF2 | |
Human SREBP2 protein (amino acids 48-403). [UniProtNo. Q12772] | |
Primary | |
Recognizes the precursor and mature forms of human SREBP2. | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1D2 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
BHLHD2, bHLHd2sterol regulatory element-binding protein 2, Class D basic helix-loop-helix protein 2, SREBP2, SREBP2SREBP-2, sterol regulatory element binding transcription factor 2, Sterol regulatory element-binding transcription factor 2 | |
Mouse | |
0.1 mL | |
6721 | |
Human, Chicken | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction