Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SREBP2 Antibody (SREBP2/1579) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308527
Description
SREBP2 Monoclonal specifically detects SREBP2 in Human samples. It is validated for ELISA.Specifications
SREBP2 | |
Monoclonal | |
1 mg/mL | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
6721 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |
ELISA | |
SREBP2/1579 | |
Unconjugated | |
BHLHD2, bHLHd2sterol regulatory element-binding protein 2, Class D basic helix-loop-helix protein 2, SREBP2, SREBP2SREBP-2, sterol regulatory element binding transcription factor 2, Sterol regulatory element-binding transcription factor 2 | |
Recombinant full-length human SREBP2 protein | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction