Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SREBP2 Mouse anti-Human, Clone: SREBP2/1579, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | SREBP2 |
---|---|
Clone | SREBP2/1579 |
Concentration | 0.2 mg/ml |
Dilution | ELISA |
Applications | ELISA |
Description
SREBP2 Monoclonal specifically detects SREBP2 in Human samples. It is validated for ELISA.Specifications
SREBP2 | |
0.2 mg/ml | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
BHLHD2, bHLHd2sterol regulatory element-binding protein 2, Class D basic helix-loop-helix protein 2, SREBP2, SREBP2SREBP-2, sterol regulatory element binding transcription factor 2, Sterol regulatory element-binding transcription factor 2 | |
Recombinant full-length human SREBP2 protein | |
Primary | |
Store at 4°C. |
SREBP2/1579 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
6721 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title