Learn More
SREK1/SFRS12 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305104AT
 
            Description
The recommended shelf life for this product is 1 year from date of receipt.
SFRS12 belongs to the superfamily of serine/arginine-rich (SR) splicing factors. It modulates splice site selection by regulating the activities of other SR proteins (Barnard et al., 2002 [PubMed 11991645]).Specifications
| SREK1/SFRS12 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q8WXA9 | |
| SREK1 | |
| Between 458 and 508 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
| Western Blot | |
| Unconjugated | |
| SREK1 | |
| SFRS12, SRrp508, SRrp86, serine-arginine-rich splicing regulatory protein 508, serine-arginine-rich-splicing regulatory protein 86, serine/arginine-rich-splicing regulatory protein 86, splicing factor, arginine/serine-rich 12, splicing regulatory protein 508 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140890 | |
| 4° C | |
| Liquid | 
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
