Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SREK1IP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566536
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566536 Supplier Invitrogen™ Supplier No. PA566536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EENEELNKLQA LQEKRINEEE EKKKEKSKEK IKLKKKRKRS Y Highest antigen sequence identity to the following orthologs - mouse 95%, rat 93%.

Possible splicing regulator involved in the control of cellular survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SREK1IP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SREK1IP1
Gene Accession No. Q8N9Q2
Gene Alias p18 splicing regulatory protein; P18SRP; protein SFRS12IP1; protein SREK1IP1; SFRS12-interacting protein 1; SFRS12IP1; Splicing regulatory protein of 18 kDa; SREK1 interacting protein 1; SREK1-interacting protein 1; SREK1IP1
Gene Symbols SREK1IP1
Host Species Rabbit
Immunogen Recombinant Human SREK1IP1. Recombinant protein control fragment (Product #RP-107199).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285672
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.