Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRF Monoclonal Antibody (2C5), Active Motif™

Rat Monoclonal Antibody
$156.65 - $638.65
Specifications
Antigen | SRF |
---|---|
Clone | 2C5 |
Concentration | 1 mg/mL |
Applications | ChIP Assay, ChIP sequencing (ChIP-seq) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3362
![]() ![]() |
Active Motif
61386 |
10 μg |
Each for $156.65
|
|
|||||
50-199-3361
![]() ![]() |
Active Motif
61385 |
100 μg |
Each for $638.65
|
|
|||||
Description
Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation. SRF-binding sites are also constitutive promotor elements in many muscle-specific promotors. SFR is required for cardiac differentiation and maturation.Specifications
SRF | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
AW049942; AW240594; BOS_21550; c-fos serum response element-binding transcription factor; LOC100354695; MCM1; minichromosome maintenance 1 homolog; OTTHUMP00000039820; RGD1559787; Serum response factor; serum response factor (c-fos serum response element-binding transcription factor); Srf | |
SRF | |
IgG2a | |
Protein G | |
Antibody |
2C5 | |
ChIP Assay, ChIP sequencing (ChIP-seq) | |
Unconjugated | |
Rat | |
Human, Mouse | |
P11831, Q9JM73 | |
20807, 6722 | |
This antibody was raised against a full-length protein corresponding to mouse SRF. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
SRF |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title