Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRF Mouse anti-Human, Clone: 1G8F4, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 667421IG150UL
Description
Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation.Specifications
| SRF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11831 | |
| SRF | |
| SRF Fusion Protein Ag10386 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1G8F4 | |
| Unconjugated | |
| SRF | |
| MCM1, Serum response factor, SRF | |
| Mouse | |
| Protein A | |
| RUO | |
| 6722 | |
| -20°C | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction