Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519176
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519176 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519176 Supplier Invitrogen™ Supplier No. PA519176
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation. SRF-binding sites are also constitutive promotor elements in many muscle-specific promotors. SFR is required for cardiac differentiation and maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRF
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SRF
Gene Accession No. P11831
Gene Alias AW049942; AW240594; c-fos serum response element-binding transcription factor; MCM1; minichromosome maintenance 1 homolog; OTTHUMP00000039820; RGD1559787; Serum response factor; serum response factor (c-fos serum response element-binding transcription factor); Srf
Gene Symbols SRF
Host Species Goat
Immunogen Synthetic peptide sequence (QASPSRDSSTDLTQ) corresponding to the internal amino acids of SRF (aa 368-382).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6722
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.