Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRFBP1 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP152655G
Description
SRFBP1 Polyclonal antibody specifically detects SRFBP1 in Human samples. It is validated for Western BlotSpecifications
SRFBP1 | |
Polyclonal | |
Western Blot | |
BUD22, BUD22 homolog, FLJ25286, p49, p49/STRAP, Rlb1, serum response factor binding protein 1, serum response factor-binding protein 1, SRF-dependent transcription regulation associated protein, SRF-dependent transcription regulation-associated protein, STRAP | |
The immunogen for this product maps to a region between residue 379 and 429 of human SRF-Dependent Transcription Regulation-Associated Protein using the numbering given in entry NP_689759.2 (GeneID 153443). | |
0.1 mL | |
Cardiovascular Biology | |
153443 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction