Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRFBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543637
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543637 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543637 Supplier Invitrogen™ Supplier No. PA543637
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LKSKKGTEDA LLKNQRRAQR LLEEIHAMKE LKPDIVTKSA LGDDINFEKI Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.

SRFBP1 may be involved in regulating transcriptional activation of cardiac genes during the aging process. It may play a role in biosynthesis and/or processing of SLC2A4 in adipose cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRFBP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Srfbp1
Gene Accession No. Q8NEF9
Gene Alias 2810036K01Rik; BUD22; BUD22 homolog; FLJ25286; P49; p49/STRAP; RGD:1359398}; Rlb1; serum response factor binding protein 1; serum response factor-binding protein 1; SRFBP1; srfbp1 {ECO:0000312; SRF-dependent transcription regulation associated protein; SRF-dependent transcription regulation-associated protein; STRAP
Gene Symbols Srfbp1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SRFBP1 (aa 38-87).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153443
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.