Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRG11/Spata17 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984272
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4272 Supplier Bioss Supplier No. BS1899R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPATA17 contains 3 IQ domains. The function of the SPATA17 protein is not known.

Specifications

Antigen SRG11/Spata17
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SPATA17
Gene Accession No. Q96L03
Gene Alias IQ motif containing H; IQCH; LOC100342220; MSRG11; MSRG-11; RP11-144C20.1; SPATA17; spermatogenesis associated 17; Spermatogenesis-associated protein 17; spermatogenesis-related protein 11
Gene Symbols SPATA17
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SRG11.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128153
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.