Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRG5/SPATA12 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984271
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4271 Supplier Bioss Supplier No. BS1898R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is expressed primarily in testis and may play a role in testicular development and spermatogenesis. The encoded protein may be upregulated in response to ultraviolet-C radiation. [provided by RefSeq, Dec 2015].

Specifications

Antigen SRG5/SPATA12
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SPATA12
Gene Accession No. Q7Z6I5
Gene Alias SPATA12; spermatogenesis associated 12; Spermatogenesis-associated protein 12; Spermatogenesis-related protein 5; SRG5; testicular tissue protein Li 184
Gene Symbols SPATA12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SPATA12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353324
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.