Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRGAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550832
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550832 Supplier Invitrogen™ Supplier No. PA550832
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SRGAP1 protein.

The protein encoded by this gene is a GTPase activator, working with the GTPase CDC42 to negatively regulate neuronal migration. The encoded protein interacts with the transmembrane receptor ROBO1 to inactivate CDC42.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRGAP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SRGAP1
Gene Accession No. Q7Z6B7, Q91Z69
Gene Alias 4930572H05Rik; Arhgap13; KIAA1304; NMTC2; RGD1566260; Rho GTPase activating protein 13; rho GTPase-activating protein 13; SLIT-ROBO Rho GTPase activating protein 1; SLIT-ROBO Rho GTPase-activating protein 1; Srgap1
Gene Symbols SRGAP1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human SLIT-ROBO Rho GTPase activating protein 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117600, 57522
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.