Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRGAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555793
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555793 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555793 Supplier Invitrogen™ Supplier No. PA555793
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEPLKTSPVV APTSEPSSPL HTQLLKDPEP AFQRSASTAG DIACAFRPVK SVKMAAPVKP PATRPKPTVF PKTNATSPGV NSSTSPQST Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.

This locus encodes a member of the SLIT-ROBO Rho GTPase activating protein family. The encoded protein stimulates GTPase activity of Rac1, and plays a role in cortical neuron development. This locus has several paralogs on human chromosome 1 resulting from segmental duplication. While this locus itself is conserved among various species, the paralogs are found only in the genus Homo, and not in the genomes of non-human great apes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRGAP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRGAP2
Gene Accession No. O75044
Gene Alias 9930124L22Rik; AI448945; ARHGAP34; FBP2; Fbp27; FBP-27; FNBP2; formin binding protein 2; formin-binding protein 2; Formin-binding protein 27; KIAA0456; RGD1566016; rho GTPase-activating protein 34; SLIT-ROBO GAP2; SLIT-ROBO Rho GTPase activating protein 2; SLIT-ROBO Rho GTPase-activating protein 2; Srgap2; SRGAP2A; srGAP3
Gene Symbols SRGAP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SRGAP2. Recombinant protein control fragment (Product #RP-94434).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23380
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.