Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRGAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578445
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578445 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578445 Supplier Invitrogen™ Supplier No. PA578445
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2, mouse brain, rat brain Predicted Reactivity: Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This locus encodes a member of the SLIT-ROBO Rho GTPase activating protein family. The encoded protein stimulates GTPase activity of Rac1, and plays a role in cortical neuron development. This locus has several paralogs on human chromosome 1 resulting from segmental duplication. While this locus itself is conserved among various species, the paralogs are found only in the genus Homo, and not in the genomes of non-human great apes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRGAP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SRGAP2
Gene Accession No. D4A208, O75044, Q91Z67
Gene Alias 9930124L22Rik; AI448945; ARHGAP34; FBP2; Fbp27; FBP-27; FNBP2; formin binding protein 2; formin-binding protein 2; Formin-binding protein 27; KIAA0456; RGD1566016; rho GTPase-activating protein 34; SLIT-ROBO GAP2; SLIT-ROBO Rho GTPase activating protein 2; SLIT-ROBO Rho GTPase-activating protein 2; Srgap2; SRGAP2A; srGAP3
Gene Symbols SRGAP2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human SRGAP2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14270, 23380, 360840
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.