Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRGAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557890
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557890 Supplier Invitrogen™ Supplier No. PA557890
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKNPPGPVSS EPASPLHTIV IRDPDAAMRR SSSSSTEMMT TFKPALSARL AGAQLRPPPM RPVRPVVQHR S Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

GTPase-activating protein for RAC1 and perhaps Cdc42, but not for RhoA small GTPase. May attenuate RAC1 signaling in neurons. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRGAP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRGAP3
Gene Accession No. O43295
Gene Alias AI452337; ARHGAP14; brain stress early protein Gbi; D130026O08Rik; Gbi; Kiaa0411; KIAA1156; MEGAP; mental disorder-associated GAP; mKIAA0411; RGD1563152; rho GTPase activating protein 14; rho GTPase-activating protein 14; SLIT-ROBO Rho GTPase activating protein 2; SLIT-ROBO Rho GTPase activating protein 3; SLIT-ROBO Rho GTPase-activating protein 3; SRGAP2; srGAP3; WAVE-associated Rac GTPase activating protein; WAVE-associated Rac GTPase-activating protein; WRP
Gene Symbols SRGAP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SRGAP3. Recombinant protein control fragment (Product #RP-96476).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9901
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.