Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRI Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300310JF549
Description
SRI Polyclonal antibody specifically detects SRI in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
SRI | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
calcium binding protein amplified in mutlidrug-resistant cells, CP22, CP-22, FLJ26259,22 kDa protein, H_RG167B05.1, SCN, sorcin, V19 | |
Produced in rabbits immunized with purified, recombinant Human SRI (Accession#: P30626; Met1-Val198) | |
0.1 mL | |
Cardiovascular Biology | |
6717 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction