Learn More
Invitrogen™ SRM Monoclonal Antibody (2C1)

Description
Peptide Sequence: LCCQGECQWL HLDLIKEMRQ FCQSLFPVVA YAYCTIPTYP SGQIGFMLCS KNPSTNFQEP VQPLTQQQVA QMQLKYYNSD VHRAAFVLPE FARKALNDV.
Specifications
Specifications
| Antigen | SRM |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2C1 |
| Concentration | 0.32 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SRM |
| Gene Accession No. | P19623 |
| Gene Alias | AA407669; PAPT; Putrescine aminopropyltransferase; SpdST; SPDSY; Spermidine synthase; spermidine synthase-1; SPS1; Srm; SRML1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.