Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553377 Supplier Invitrogen™ Supplier No. PA553377
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FKESYYQLMK TALKEDGVLC CQGECQWLHL DLIKEMRQFC QSLFPVVAYA YCTIPTYPSG QIGFMLCSKN PSTNFQEPVQ PLTQQQVAQM QLKYYNSDVH RAAFVLPEF Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

The polyamines putrescine, spermine, and spermidine are ubiquitous polycationic mediators of cell growth and differentiation. SRM synthase is one of four enzymes in the polyamine-biosynthetic pathway and carries out the final step of spermidine biosynthesis. This enzyme catalyzes the conversion of putrescine to spermidine using decarboxylated S-adenosylmethionine as the cofactor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRM
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRM
Gene Accession No. P19623
Gene Alias AA407669; PAPT; Putrescine aminopropyltransferase; SpdST; SPDSY; Spermidine synthase; spermidine synthase-1; SPS1; Srm; SRML1
Gene Symbols SRM
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SRM. Recombinant protein control fragment (Product #RP-91418).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6723
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.