Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596953
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596953 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596953 Supplier Invitrogen™ Supplier No. PA596953
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEPGPDGPAA SGPAAIREGW FRETCSLWPG QALSLQVEQL LHHRRSRYQD ILVFRSKTYG NVLVLDGVIQ CTERDEFSYQ EMIANLPLCS HPNPRKVLII GGGDGGVLRE VVKHPSVESV VQCEIDEDVI QVSKKFLPGM AIGYSSSKLT LHVGDGFEFM KQNQDAFDVI ITDSSDPMGP AESLFKESYY QLMKTALKED GVLCCQGECQ WLHLDLIKEM RQFCQSLFPV VAYAYCTIPT YPSGQIGFML CSKNPSTNFQ EPVQPLTQQQ VAQMQLKYYN SDVHRAAFVL PEFARKALND VS; Positive Samples: 22Rv1, HepG2, HeLa, Mouse liver, Mouse stomach, Mouse small intestine, Rat liver.

The polyamines putrescine, spermine, and spermidine are ubiquitous polycationic mediators of cell growth and differentiation. SRM synthase is one of four enzymes in the polyamine-biosynthetic pathway and carries out the final step of spermidine biosynthesis. This enzyme catalyzes the conversion of putrescine to spermidine using decarboxylated S-adenosylmethionine as the cofactor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRM
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SRM
Gene Accession No. P19623, Q64674
Gene Alias AA407669; PAPT; Putrescine aminopropyltransferase; SpdST; SPDSY; Spermidine synthase; spermidine synthase-1; SPS1; Srm; SRML1
Gene Symbols SRM
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-302 of human SRM (NP_0031232).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20810, 6723, 84596
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.