Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRMS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100918
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100918 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100918 Supplier Invitrogen™ Supplier No. PA5100918
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SRMS.

SRMS (Srm) is a tyrosine protein kinase (SRC sub-family) that may be involved in the proliferation and differentiation of keratinocytes. Non-receptor tyrosine-protein kinase which phosphorylates DOK1 on tyrosine residues. Also phosphorylates KHDRBS1/SAM68 and VIM on tyrosine residues. Phosphorylation of KHDRBS1 is EGF-dependent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRMS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SRMS
Gene Accession No. Q9H3Y6
Gene Alias A230069J08Rik; C20orf148; dJ697K14.1; PTK70; src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites; SRM; SRMS; Tyrosine-protein kinase Srms
Gene Symbols SRMS
Host Species Rabbit
Immunogen A synthesized peptide derived from human SRMS(Accession Q9H3Y6), corresponding to amino acid residues K406-Y456.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296472, 6725
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.