Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRMS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA514790
Description
SRMS Polyclonal Antibody for Western Blot, IHC (P)
SRMS (Srm) is a tyrosine protein kinase (SRC sub-family) that may be involved in the proliferation and differentiation of keratinocytes. Non-receptor tyrosine-protein kinase which phosphorylates DOK1 on tyrosine residues. Also phosphorylates KHDRBS1/SAM68 and VIM on tyrosine residues. Phosphorylation of KHDRBS1 is EGF-dependent.
Specifications
SRMS | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9H3Y6 | |
SRMS | |
KLH conjugated synthetic peptide between 19-48 amino acids from the N-terminal region of human SRMS. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SRMS | |
A230069J08Rik; C20orf148; dJ697K14.1; PTK70; src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites; SRM; SRMS; Tyrosine-protein kinase Srms | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
6725 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction