Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRMS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572209
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572209 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572209 Supplier Invitrogen™ Supplier No. PA572209
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRMS Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

SRMS (Srm) is a tyrosine protein kinase (SRC sub-family) that may be involved in the proliferation and differentiation of keratinocytes. Non-receptor tyrosine-protein kinase which phosphorylates DOK1 on tyrosine residues. Also phosphorylates KHDRBS1/SAM68 and VIM on tyrosine residues. Phosphorylation of KHDRBS1 is EGF-dependent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRMS
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SRMS
Gene Accession No. Q62270
Gene Alias A230069J08Rik; C20orf148; dJ697K14.1; PTK70; src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites; SRM; SRMS; Tyrosine-protein kinase Srms
Gene Symbols SRMS
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 113-147 amino acids from the Central region of Mouse Srms.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20811
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.