Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRP14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541768
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541768 Supplier Invitrogen™ Supplier No. PA541768
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KPIPKKGTVE GFEPADNKCL LRATDGKKKI STVVSSKEVN KFQMAYSNLL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 86%.

Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRP14
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Srp14
Gene Accession No. P37108
Gene Alias (homologous Alu RNA binding protein); 14kDa; 18 kDa Alu RNA-binding protein; ALURBP; AW536328; homologous Alu RNA binding protein; homologous Alu RNA-binding protein; hypothetical protein LOC619260; signal recognition particle 14; signal recognition particle 14 kDa protein; signal recognition particle 14kDa (homologous Alu RNA binding protein); SRP14; zgc:112290
Gene Symbols Srp14
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SRP14 (aa 38-87).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6727
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.