Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRP14 Rabbit anti-Human, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 115281AP150UL
Description
Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.Specifications
| SRP14 | |
| Polyclonal | |
| Unconjugated | |
| SRP14 | |
| 18 kDa Alu RNA binding protein, ALURBP, SRP14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 296076, 6727 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P37108 | |
| SRP14 | |
| SRP14 Fusion Protein Ag2099 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction