Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRP19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541769 Supplier Invitrogen™ Supplier No. PA541769
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LCLVQFPSRK SVMLYAAEMI PKLKTRTQKT GGADQSLQQG EGSKKGKGKK Sequence homology: Cow: 93%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 92%; Pig: 93%; Rabbit: 85%; Rat: 92%; Zebrafish: 77%.

SRP19 belongs to the SRP19 family. It is signal-recognition-particle assembly and binds directly to 7S RNA and mediates binding of the 54 kDa subunit of the SRP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRP19
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SRP19
Gene Accession No. P09132
Gene Alias 2310020D23Rik; hypothetical protein LOC514960; signal recognition particle; signal recognition particle 19; signal recognition particle 19 kDa protein; signal recognition particle 19kD; signal recognition particle 19kDa; SRP19; zgc:63961
Gene Symbols SRP19
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SRP19.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6728
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.