Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRP19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593197
Description
Immunogen sequence: MACAAARSPA DQDRFICIYP AYLNNKKTIA EGRRIPISKA VENPTATEIQ DVCSAVGLNV FLEKNKMYSR EWNRDVQYRG RVRVQLKQED GSLCLVQFPS RKSVMLYAAE MIPKLKTRTQ KTGGADQSLQ QGEGSKKGKG KKKK; Positive Samples: HepG2, MCF7; Cellular Location: Cytoplasm.
SRP19 belongs to the SRP19 family. It is signal-recognition-particle assembly and binds directly to 7S RNA and mediates binding of the 54 kDa subunit of the SRP.
Specifications
SRP19 | |
Polyclonal | |
Unconjugated | |
SRP19 | |
2310020D23Rik; hypothetical protein LOC514960; signal recognition particle; signal recognition particle 19; signal recognition particle 19 kDa protein; signal recognition particle 19kD; signal recognition particle 19kDa; SRP19; zgc:63961 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
291685, 66384, 6728 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09132, Q9D7A6 | |
SRP19 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-144 of human SRP19 (NP_0031261). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction