Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRP54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566449 Supplier Invitrogen™ Supplier No. PA566449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VIIASEGVEKF KNENFEIIIV DTSGRHKQED SLFEEMLQVA NAIQPDNIVY VMDASIGQAC EAQAKAFKDK VDVASVIVT Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

Binds to the signal sequence of presecretory protein when they emerge from the ribosomes and transfers them to TRAM (translocating chain-associating membrane protein).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRP54
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRP54
Gene Accession No. P61011
Gene Alias 54K subunit of signal recognition particle; 54kDa; signal recognition particle 54; signal recognition particle 54 kDa protein; signal recognition particle 54A; signal recognition particle 54kD; signal recognition particle 54kDa; SRP54; Srp54a
Gene Symbols SRP54
Host Species Rabbit
Immunogen Recombinant Human SRP54. Recombinant protein control fragment (Product #RP-105221).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6729
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.